Skip to Content
Merck
All Photos(1)

Key Documents

212989

Sigma-Aldrich

Bromochlorophenol Blue sodium salt

Synonym(s):

3′,3′′-Dibromo-5′,5′′-dichlorophenolsulfonephthalein, 3′,3′′-Dibromo-5′,5′′-dichlorophenolsulfonephthalein sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H9Br2Cl2NaO5S
CAS Number:
Molecular Weight:
603.04
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

visual transition interval

3.0-4.6, yellow to blue

λmax

590 nm

SMILES string

[Na+].Oc1c(Cl)cc(cc1Br)\C(=C2\C=C(Cl)C(=O)C(Br)=C2)c3ccccc3S([O-])(=O)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service