Skip to Content
Merck
All Photos(1)

Documents

07586

Supelco

Methyl trans-vaccenate

analytical standard

Synonym(s):

trans-11-Octadecenoic acid methyl ester, Methyl trans-11-octadecenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H36O2
CAS Number:
Molecular Weight:
296.49
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

[H]\C(CCCCCC)=C(\[H])CCCCCCCCCC(=O)OC

InChI

1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h8-9H,3-7,10-18H2,1-2H3/b9-8+

InChI key

PVVODBCDJBGMJL-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl trans-vaccenate belongs to the class of unsaturated fatty acid methyl esters. These unsaturated fatty acids are easily oxidized. Flush open containers with argon or nitrogen and store at -20°C, in dark.

Application

Methyl trans-vaccenate may be used as an analytical reference standard in the following:
  • Determination of the analyte subjected to cellular fatty acid profile analysis by gas chromatography (GC) and GC-mass spectrometry (MS).
  • Quantification of the analyte via determining the milk fatty acid profile in lactating ewes exposed to high ambient temperature using gas chromatography with flame ionization detection.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of solar radiation and flaxseed supplementation on milk production and fatty acid profile of lactating ewes under high ambient temperature
Caroprese M, et al.
Journal of Dairy Science, 94(8), 3856-3867 (2011)
Advanced gas chromatographic?mass spectrometric studies for identification of the cellular octadecenoate isomers and changes of fatty acid composition induced by ethanol stress in Escherichia coli and Escherichia coli O157: H7
Ku L-K , et al.
Journal of Food Protection, 70(3), 616-622 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service