Skip to Content
Merck
All Photos(1)

Documents

WBPRPKL01

Milli-Q

Pack Holder for PrePak L Pretreatment Pack

Securely supports the PrePak L Pretreatment pack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
JA.13

packaging

pkg of 1 unit

H × W × D

795 mm (31.3 in.) × 234 mm (9.2 in.) × 192 mm (7.6 in.)

compatibility

for use with Elix®
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
for use with RiOs

shipped in

ambient

General description

The PrePak L wall mounting bracket is a water purification accessory designed to support the PrePak L pretreatment pack for a leak-free and secure fitting.
The unit includes wall bracket, fittings, tubing, and pressure gauges required for the installation and operation of the PrePak L pretreatment pack that protects Elix® and RiOs water purification systems.

Features and Benefits

Tight and leak-free fitting of PrePak L Pretreatment pack

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service