MXQUANK01-M
Milliflex® Quantum standard kit
Easy-to-use, non-destructive, fluorescent staining-based system for faster microbial detection
Synonym(s):
Fluorescence-based rapid detection system
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116129
eCl@ss:
32160606
NACRES:
NB.79
Recommended Products
manufacturer/tradename
Milliflex®
input
sample(s) (in process)
water
sample(s) (final product)
sample(s) (raw material)
sample(s) (environmental)
application(s)
pharmaceutical
General description
Milliflex® Quantum Standard Kit is a fluorescence-based technology for the quantitative and rapid detection of microorganisms in filterable samples. The universal enzymatic fluorescent staining is non-destructive, allowing the identification of the contaminant following a positive result. Therefore, the Milliflex® Quantum system is a fast and reliable alternative for the detection of microorganisms.
Application
The Milliflex® Quantum Standard Kit is used for rapid bioburden testing in filtrable samples, including in-process and final product samples (pharmaceuticals and water). The product is also used in industrial QC and water monitoring.
Features and Benefits
- Non-destructive method enables reliable identification using any ID technology
- Easy-to-use system with a simple workflow requires minimal training
- Results comparable to a compendial method, enabling faster validation to benefit from rapid detection
- Economical, robust system
- Compact hardware fits on any laboratory bench
Components
1 reader
1 reader stand
1 membrane transfer tool
1 removal rack
1 camera
Installation files on USB key
1 reader stand
1 membrane transfer tool
1 removal rack
1 camera
Installation files on USB key
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service