MABF3148
Anti-Rabies Virus Phosphoprotein Antibody, clone M974
Synonym(s):
Protein P;Protein M1
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
M974, monoclonal
mol wt
calculated mol wt 33.21 kDa
observed mol wt ~44 kDa
purified by
using protein G
species reactivity
rabies virus
packaging
antibody small pack of 100 μg
technique(s)
ELISA: suitable
inhibition assay: suitable
western blot: suitable
isotype
IgG2aκ
epitope sequence
N-terminal half
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting with recombinant Rabies Virus Phosphoprotein.
Western Blotting Analysis: 1: 500,000 dilution (1 ng/mL) of this antibody detected recombinant Rabies Virus Phosphoprotein.
Tested applications
Western Blotting Analysis: A representative lot detected Rabies Virus Phosphoprotein in Western Blotting applications (Nadin-Davis, S.A., et al. (2000). J Clin Microbiol. 38(4):1397-1403).
ELISA Analysis: A representative lot detected Rabies Virus Phosphoprotein in ELISA applications (Nadin-Davis, S.A., et al. (2000). J Clin Microbiol. 38(4):1397-1403).
Inhibition Assay: A representative lot of this antibody inhibited binding of Rabies virus phosphoprotein in a competitive binding assay. (Nadin-Davis, S.A. , et al. (2000). J Clin Microbiol. 38(4):1397-403).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service