Skip to Content
Merck
All Photos(5)

Key Documents

MAB5232

Sigma-Aldrich

Anti-Presenilin-1 Antibody, loop, a.a. 263-378, CT, clone PS1-loop

ascites fluid, clone PS1-loop, Chemicon®

Synonym(s):

Anti-ACNINV3, Anti-AD3, Anti-FAD, Anti-PS1, Anti-S182

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

PS1-loop, monoclonal

species reactivity

rat, primate, mouse, human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PSEN1(5663)

General description

Presenilin is a probable catalytic subunit of the gamma-secretase complex, an endoprotease complex that catalyzes the intramembrane cleavage of integral membrane proteins such as Notch receptors and APP (beta-amyloid precursor protein). Presenilin requires the other members of the gamma-secretase complex to have a protease activity. It may play a role in intracellular signaling and gene expression or in linking chromatin to the nuclear membrane. The gamma-secretase complex stimulates cell-cell adhesion through its association with the E-cadherin/catenin complex. Under conditions of apoptosis or calcium influx, it cleaves E-cadherin promoting the disassembly of the E-cadherin/catenin complex and increasing the pool of cytoplasmic beta-catenin, thus negatively regulating Wnt signaling. Futher researcher may implicate the complex in hematopoiesis. Defects in PSEN1 have been implicated in early-onset Alzheimer disease type 3 (AD3) and late-onset Alzheimer disease (LOAD).

Specificity

Presenilin-1, loop

Immunogen

Epitope: C-terminus
Presenilin-1, loop fusion protein (amino acids 263-378), human

Application

Anti-Presenilin-1 Antibody, loop, a.a. 263-378, C-terminus, clone PS1-loop detects level of Presenilin-1 & has been published & validated for use in IC, IH(P), IP & WB.
Immunocytochemistry:
A previous lot of this antibody was used in IC.Acetone or methanol fixed cells; or lightly fixed 2.5-3% PFA (5-10′ RT).

Immunohistochemistry:
A previous lot was used on 3% paraformaldehyde fixed tissue. Works on paraffin embedded tissue sections if fixed with PLP, acetone or other light fixations; 10% formalin fixation is untested.

Immunoprecipitation:
A previous lot of this antibody was used in IP.

Western blot:
Recognizes a protein of approximately 18-20 kD, and occasionally there is a doublet (the top band of which is phosphorylated).

Optimal working dilutions must be determined by the end user
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Routinely evaluated by Western Blot on PC12 lysates.

Western Blot Analysis:
1:500 dilution of this lot detected PRESENILIN-1 on 10 μg of PC12 lysates.

Target description

18-20 kDa

Physical form

Mouse monoclonal IgG1 ascites with no preservatives.
Unpurified

Storage and Stability

Stable for 6 months at -20ºC in undiluted aliquots from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuhiro Teranishi et al.
Journal of cellular and molecular medicine, 14(11), 2675-2686 (2009-09-17)
γ-Secretase is a transmembrane protease complex responsible for the processing of a multitude of type 1 transmembrane proteins, including amyloid precursor protein (APP) and Notch. A functional complex is dependent on the assembly of four proteins: presenilin (PS), nicastrin, Aph-1
Presenilin-1 regulates induction of hypoxia inducible factor-1?: altered activation by a mutation associated with familial Alzheimer's disease.
De Gasperi, R; Sosa, MA; Dracheva, S; Elder, GA
Mol. Neurodegener. null
Xiao-Juan Cheng et al.
PloS one, 10(6), e0130432-e0130432 (2015-06-16)
Recent studies suggest that high-salt diet is associated with cognitive decline in human and mouse. The fact that genetic factors account for less than 50% cases of sporadic Alzheimer's disease (AD) highlights the important contribution of environmental factors, such as
Tokiaki Yamaguchi et al.
Scientific reports, 6, 34505-34505 (2016-10-01)
Alzheimer's disease (AD) is the most prevalent form of dementia characterized by the extracellular accumulation of amyloid β (Aβ) peptides, which are produced by proteolytic cleavages of amyloid precursor protein (APP). Gangliosides are involved in AD pathophysiology including Aβ deposition
Yi Li et al.
Structure (London, England : 1993), 22(1), 125-135 (2013-11-12)
Presenilin-mediated endoproteolysis of transmembrane proteins plays a key role in physiological signaling and in the pathogenesis of Alzheimer disease and some cancers. Numerous inhibitors have been found via library screens, but their structural mechanisms remain unknown. We used several biophysical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service