Skip to Content
Merck
All Photos(1)

Key Documents

AB5818

Sigma-Aldrich

Anti-Potassium Channel Kir4.1 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
rat, mouse, human
technique(s):
western blot: suitable
citations:
8
Pricing and availability is not currently available.

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KCNJ10(3766)

Specificity

Recognizes Kir4.1. The epitope does not share homology with any other known proteins.

SPECIES REACTIVITIES: The antibody is also expected to work with mouse and human due to sequence homology. Other species have not been tested.

Immunogen

CONTROL ANTIGEN: Included free of charge with the antibody is 20 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 μL of sterile distilled water. For negative control, preincubate 1 μg of protein with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user. Synthetic peptide corresponding to amino acids 356-375 of rat, mouse or human Kir4.1 (Accession P49655).

Application

This Anti-Potassium Channel Kir4.1 Antibody is validated for use in WB for the detection of Potassium Channel Kir4.1.
Western blot: 1:200-1:600 using ECL on rat brain membranes or human platelets.

Dilutions should be made using a carrier protein such as BSA (1-3%)

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, 5% sucrose as a stabilizer and 0.025% sodium azide as a preservative. Reconstitute with 50 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for

Analysis Note

Control
Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 μL of sterile distilled water. For negative control, preincubate 1 μg of protein with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mieke Wesdorp et al.
American journal of human genetics, 103(1), 74-88 (2018-07-03)
In a Dutch consanguineous family with recessively inherited nonsyndromic hearing impairment (HI), homozygosity mapping combined with whole-exome sequencing revealed a MPZL2 homozygous truncating variant, c.72del (p.Ile24Metfs∗22). By screening a cohort of phenotype-matched subjects and a cohort of HI subjects in
Yue Yang et al.
PloS one, 6(1), e16547-e16547 (2011-02-10)
The endothelial-blood/tissue barrier is critical for maintaining tissue homeostasis. The ear harbors a unique endothelial-blood/tissue barrier which we term "blood-labyrinth-barrier". This barrier is critical for maintaining inner ear homeostasis. Disruption of the blood-labyrinth-barrier is closely associated with a number of
Zuzanna Setkowicz et al.
Journal of anatomy, 231(3), 366-379 (2017-06-10)
Astrocytes participate in neuronal development and excitability, and produce factors enhancing or suppressing inflammatory processes occurring due to neurodegenerative diseases, such as epilepsy. Seizures, in turn, trigger the release of inflammatory mediators, causing structural and functional changes in the brain.
Meritxell Espino Guarch et al.
eLife, 7 (2018-01-23)
Age-related hearing loss (ARHL) is the most common sensory deficit in the elderly. The disease has a multifactorial etiology with both environmental and genetic factors involved being largely unknown. SLC7A8/SLC3A2 heterodimer is a neutral amino acid exchanger. Here, we demonstrated
Steven A Hill et al.
eLife, 8 (2019-06-14)
Astrocytes have emerged as integral partners with neurons in regulating synapse formation and function, but the mechanisms that mediate these interactions are not well understood. Here, we show that Sonic hedgehog (Shh) signaling in mature astrocytes is required for establishing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service