Skip to Content
Merck
All Photos(2)

Key Documents

AB5166

Sigma-Aldrich

Anti-Muscarinic Acetylcholine Receptor m2 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CHRM2(1129)

Specificity

Recognizes a full-length m2 protein. It has exhibited no cross reactivity with other muscarinic proteins tested so far.

SPECIES REACTIVITIES: In Chimpanzee and gorilla the immunogen sequence is 100% identical. Orangutan (130/132), pig (122/132), and dog (122/132). Other species have not been tested.

Immunogen

GST fusion protein (Rybin, 2000) and part of i3 intercellular loop of human m2 muscarinic acetylcholine receptor (amino acids 225-356) (Accession P08172).

Application

Anti-Muscarinic Acetylcholine Receptor m2 Antibody is an antibody against Muscarinic Acetylcholine Receptor m2 for use in IH, IP & WB.
Immunohistochemistry on rat brain sections.

Western blot: 1:200 using ECL on rat brain membranes.

Immunoprecipitation (Vorobiov, 2000; Lee, 2000)

Dilutions should be made using a carrier protein such as BSA (1-3%)

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Physical form

Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.05% sodium azide as a preservative. Reconstitute with 200 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 50 μg of control antigen (lyophilized powder in phosphate buffered saline, pH 7.4, containing 5% sucrose and 0.025% sodium azide). The stock solution of the antigen can be made up using 100 μL of sterile deionized water. For positive control, in Western blot using 50 ng of protein per lane. For negative control, preincubate 5-7 μg of fusion protein with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abraham Kovoor et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 25(8), 2157-2165 (2005-02-25)
Regulator of G-protein signaling 9-2 (RGS9-2), a member of the RGS family of G GTPase accelerating proteins, is expressed specifically in the striatum, which participates in antipsychotic-induced tardive dyskinesia and in levodopa-induced dyskinesia. We report that RGS9 knock-out mice develop
Anita A Disney et al.
The Journal of comparative neurology, 507(5), 1748-1762 (2008-02-12)
Acetylcholine (ACh) is believed to underlie mechanisms of arousal and attention in mammals. ACh also has a demonstrated functional effect in visual cortex that is both diverse and profound. We have reported previously that cholinergic modulation in V1 of the
Xiaojun Wang et al.
Cell reports, 26(11), 3145-3159 (2019-03-14)
Parsing diverse nerve cells into biological types is necessary for understanding neural circuit organization. Morphology is an intuitive criterion for neuronal classification and a proxy of connectivity, but morphological diversity and variability often preclude resolving the granularity of neuron types. Combining
Tatsuya Ogura et al.
Journal of neurophysiology, 106(3), 1274-1287 (2011-06-17)
The mammalian olfactory epithelium is made up of ciliated olfactory sensory neurons (OSNs), supporting cells, basal cells, and microvillous cells. Previously, we reported that a population of nonneuronal microvillous cells expresses transient receptor potential channel M5 (TRPM5). Using transgenic mice
Zoé Christenson Wick et al.
eLife, 8 (2019-10-15)
The hippocampus, a brain region that is important for spatial navigation and episodic memory, benefits from a rich diversity of neuronal cell-types. Through the use of an intersectional genetic viral vector approach in mice, we report novel hippocampal neurons which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service