Skip to Content
Merck
All Photos(1)

Key Documents

AB15414

Sigma-Aldrich

Anti-MAP1LC3 C Antibody

serum, Chemicon®

Synonym(s):

Microtubule-associated proteins 1A/1B light chain 3C, MAP1A/1B LC3 C, MAP1 LC3, LC3 C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Autophagy is the process by which cells recycle cytoplasm and dispose of excess or defective organelles. This process is suggested to be involved development, differentiation, growth regulation and tissue remodeling in multicellular organisms. The human MAP1LC3 protein is homologous with the rat Map1LC3 and with yeast APG8. These proteins are likely to be involved in the formation of autophagosomal vacuoles (autophagosomes).

Specificity

MAP1LC3 C [Microtubule-associated proteins 1A/1B light chain 3C, MAP1A/1B LC3 C, MAP1 LC3, LC3 C].

Immunogen

Synthetic peptide corresponding to a region within the N-terminus of human MAP1LC3 C protein.

Application

Research Category
Neuroscience
Research Sub Category
Neurofilament & Neuron Metabolism
This Anti-MAP1LC3 C Antibody is validated for use in WB, IC, IH for the detection of MAP1LC3 C.
Western blot: 1:100-1:1,000
Immunocytochemistry: 1:100-1:1,000
Immunohistochemistry: 1:100-1:1,000
Optimal working dilutions must be determined by end user.

Physical form

Lyophilized. Reconstitute with 50 uL of sterile distilled water. Centrifuge to remove any insoluble material. Contains no preservative.
Serum

Storage and Stability

Maintain lyophilized material frozen at -20°C to -70°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. Glycerol (1:1, ACS or better grade) can be added for additional stability.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service