Skip to Content
Merck
All Photos(1)

Key Documents

8.14964

Sigma-Aldrich

1-Bromo-4-(trifluoromethoxy)benzene

for synthesis

Synonym(s):

1-Bromo-4-(trifluoromethoxy)benzene, p-Trifluoromethoxy bromo benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4BrF3O
CAS Number:
Molecular Weight:
241.01
MDL number:
UNSPSC Code:
12352112
EC Index Number:
206-979-2
Pricing and availability is not currently available.

vapor pressure

20 hPa ( 55 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

>2500 mg/kg LD50, oral (Rat)

bp

91-92 °C/100 hPa

solubility

11.7 mg/L

density

1.62 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H4BrF3O/c8-5-1-3-6(4-2-5)12-7(9,10)11/h1-4H

InChI key

SEAOBYFQWJFORM-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service