Skip to Content
Merck
All Photos(1)

Key Documents

8.04016

Sigma-Aldrich

Furfurylamine

for synthesis

Synonym(s):

Furfurylamine, (2-Furylmethyl)amine, 2-Aminomethylfuran, 2-Furanmethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO
CAS Number:
Molecular Weight:
97.12
MDL number:
UNSPSC Code:
12352116
EC Index Number:
210-536-9
Pricing and availability is not currently available.

vapor pressure

4 hPa ( 20 °C)

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

350 °C (DIN 51794)

potency

200-2000 mg/kg LD50, oral (Rat)
100 mg/kg LD50, skin (Rat)

expl. lim.

1.8 % (v/v)

pH

11.6 (20 °C, 100 g/L in H2O)

bp

145-146 °C/1013 hPa

mp

-70 °C

transition temp

flash point 46 °C (DIN 51755 Part 1)

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C5H7NO/c6-4-5-2-1-3-7-5/h1-3H,4,6H2

InChI key

DDRPCXLAQZKBJP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.049 - 1.052
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - DIN 51755 Part 1

Flash Point(C)

46 °C - DIN 51755 Part 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service