Skip to Content
Merck
All Photos(1)

Documents

5.30654

Sigma-Aldrich

c-Rel Inhibitor, IT-603

Synonym(s):

c-Rel Inhibitor, IT-603, (5 Z)-5-((5-Bromo-2-hydroxy-3-methoxyphenyl)methylidene)-2-sulfanylideneimidazolidin-4-one, (5 Z)-5-(5-Bromo-2-hydroxy-3-methoxybenzylidene)-2-thioxo-4-imidazolidinone, IT603, (5Z)-5-((5-Bromo-2-hydroxy-3-methoxyphenyl)methylidene)-2-sulfanylideneimidazolidin-4-one, (5Z)-5-(5-Bromo-2-hydroxy-3-methoxybenzylidene)-2-thioxo-4-imidazolidinone, IT603

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9BrN2O3S
CAS Number:
Molecular Weight:
329.17
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

COC1=CC(=CC(=C1O)C=C2C(=O)NC(=S)N2)Br

General description

A cell-permeable benzylidene-thioxo-imidazolidinone compund that directly and reversibly binds to c-Rel to change its conformation and blocks its DNA binding and transcriptional activity (IC50 = 3 µM in EMSA). Shows good selectivity over Oct1 and AP1. Reduces IL-2 expression in activated T cells without affecting their viability. Induces growth arrest in diffuse large B-cell lymphoma (DLBCL) cell line Ly3 (IC50 = 18 uM). Shown to reduce alloactivation in T cells and diminish graft-versus-host disease (GVHD), but does not impair TCR-mediated killing. Improves the survival rate of lethally irradiated BABL/c mice transplanted with T cell depleted bone marrow cells from C57BL/6 mice (~24 mg/kg, i.p, daily).
A cell-permeable benzylidene-thioxo-imidazolidinone compund that directly and reversibly binds to c-Rel to change its conformation and blocks its DNA binding and transcriptional activity (IC50 = 3 µM in EMSA). Shows good selectivity over Oct1 and AP1. Reduces IL-2 expression in activated T cells without affecting their viability. Induces growth arrest in diffuse large B-cell lymphoma (DLBCL) cell line Ly3 (IC50 = 18 uM). Shown to reduce alloactivation in T cells and diminish graft-versus-host disease (GVHD), but does not impair TCR-mediated killing. Improves the survival rate of lethally irradiated BABL/c mice transplanted with T cell depleted bone marrow cells from C57BL/6 mice (~24 mg/kg, i.p, daily).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
c-Rel
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Shono, Y., et al. 2014. Cancer Discovery4, 578.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service