Skip to Content
Merck
All Photos(1)

Key Documents

5.09164

Sigma-Aldrich

Windorphen

Synonym(s):

Windorphen, Wnt inhibitor dorsalizing, Histone Acetyltransferase Inhibitor XII, ( E) & ( Z)-3-Chloro-2,3- bis(4-methoxyphenyl)acrylaldehyde, WD, HAT Inhibitor XII, Wnt Pathway Inhibitor XXII, Wnt inhibitor dorsalizing, Histone Acetyltransferase Inhibitor XII, (E) & (Z)-3-Chloro-2,3-bis(4-methoxyphenyl)acrylaldehyde, WD, HAT Inhibitor XII, Wnt Pathway Inhibitor XXII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15ClO3
CAS Number:
Molecular Weight:
302.75
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

Cl\C(=C(/c2ccc(cc2)OC)\C=O)\c1ccc(cc1)OC

InChI

1S/C17H15ClO3/c1-20-14-7-3-12(4-8-14)16(11-19)17(18)13-5-9-15(21-2)10-6-13/h3-11H,1-2H3/b17-16-

InChI key

VNRALGZMXHFBPG-MSUUIHNZSA-N

General description

A cell-permeable bis(4-methoxyphenyl)chloroacrylaldehyde whose (Z)-isomer (isomeric content 12-15%) acts as a p300-selective histone acetytransferase (HAT) inhibitor (IC50 = 4.2 µM) and selectively disrupts β-catenin association with p300, but not CBP or LEF1/TCF4, exhibiting much reduced potency against KAT5, CBP, MYST4, MYST2 (IC50 = 38.2, 51.3, 59.5, 62.2 µM, respectively) and little or no inhibitory efficacy toward GCN5 and PCAF HAT activity (IC50 >100 µM). Shown to preferentially suppress human and mouse β-catenin-, zebrafish β-catenin-1-, but not zebrafish β-catenin-2-, mediated transcription activity in various reporter assays (20 µM) without affecting β-catenin activation or nuclear translocation. Induces apoptosis in Wnt signaling-dependent cancer cultures (IC50 in µM = 15.0/SW480, 19.2/RKO, 21.8/DU135, and 19.0/PC3; 72 h) in vitro without affecting the viability of Wnt-independent human lung cander cell line H460 (up to 200 µM & 72 h) and selectively abrogates Wnt signaling in ventral & lateral regions, but not within dorsal organizer, in 5.3 hpf epiboly stage zebrafish embryos in vivo, indicating that the Zebrafish β-catenin-1 & -2 regulate separate Wnt signaling events during zebrafish embryo development by associating with distinct binding partners. A great complement to the selective β-catenin-CBP interaction blocker ICG-001 (Cat. no. 504712). Pure (E)-isomer is available separately (Cat. no. xxxxxx) as a negative control.
The racemic mixture of a cell-permeable (4-methoxyphenyl)chloroacrylaldehyde compound whose (Z)-isomer (isomer content 12-15%) acts as a p300-selective histone acetytransferase (HAT) inhibitor (IC50 = 4.2 µM using racemic mixture with 13% Z-isomer) and selectively disrupts β-catenin association with p300, but not CBP or LEF1/TCF4, exhibiting much reduced potency against KAT5, CBP, MYST4, MYST2 (IC50 ≥38 µM) and little or no inhibitory efficacy toward GCN5 and PCAF HAT activity (IC50 >100 µM). Shown to selectively prevents human & mouse β-catenin-, zebrafish β-catenin-1-, but not zebrafish β-catenin-2-, mediated transcription activity without affecting β-catenin activation or nuclear translocation. Induces apoptosis in Wnt signaling-dependent cancer cultures (IC50 15 to 22 µM; 72 h) in vitro without affecting the viability of Wnt-independent H460 (up to 200 µM & 72 h) and selectively abrogates Wnt signaling in ventral & lateral regions, but not within dorsal organizer, in 5.3 hpf epiboly stage zebrafish embryos in vivo. A great complement to the selective β-catenin-CBP interaction blocker ICG-001 (Cat. no. 504712). (E)-isomer is available separately (Cat. no. 509166) as negative control.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Use only fresh DMSO for reconstitution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service