Skip to Content
Merck
All Photos(1)

Key Documents

480458

Sigma-Aldrich

NF-κB Activation Inhibitor X, Methysticin

Synonym(s):

NF-κB Activation Inhibitor X, Methysticin, (6R)-6-[(1E)-2-(1,3-Benzodioxol-5-yl)ethenyl]-5,6- dihydro-4-methoxy-2H-pyran-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O5
CAS Number:
Molecular Weight:
274.27
MDL number:
UNSPSC Code:
12352200

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C15H14O5/c1-17-12-7-11(20-15(16)8-12)4-2-10-3-5-13-14(6-10)19-9-18-13/h2-6,8,11H,7,9H2,1H3/b4-2+

InChI key

GTEXBOVBADJOQH-DUXPYHPUSA-N

General description

Methysticin has been identified from kava to have potent NF-κB inhibitory activity and minimum toxicity. It is shown to inhibit NF-κB (IC50 = 0.19 µg/ml) 100 times more potently than traditional kava in A549 cells in an in vitro luciferase-based assay. It is also reported to suppress the elevation of IKKα, p65, and COX-2 and the degradation of IκBα in A549 cultures stimulated with TNF-alpha; through Western Blot analysis.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Shaik, A.A., et al. 2009. Bioorg Med Chem Lett.19, 5732.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service