Skip to Content
Merck
All Photos(2)

Documents

454586

Sigma-Aldrich

NMDAR2C/2D Inhibitor, DQP-1105

The NMDAR2C/2D Inhibitor, DQP-1105 controls the biological activity of NMDAR2C/2D. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

NMDAR2C/2D Inhibitor, DQP-1105, NMDA Antagonist IV, DQP-1105

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H24BrN3O4
CAS Number:
Molecular Weight:
558.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow-white

solubility

DMSO: 10 mg/mL, pale yellow

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(O)CCC(N1N=C(C2=C(C3=CC=CC=C3)C4=C(NC2=O)C=CC(C)=C4)CC1C5=CC=C(Br)C=C5)=O

General description

A dihydroquinilone-pyrazoline NMDA antagonist, which is selective for NMDA receptors that are comprised of recombinant subunits GluN1/GluN2C (IC50 = 5.4 µM) and GluN1/GluN2D (2.2 µM) over GluN1/GluN2B (206 µM) and GluN1/ClucN2A (undetectable) in Xenopus oocytes. Similar selectivity is observed in whole-cell HEK patch-clamp assays with IC50 values for GluN2C and GluN2D that are at least 50-fold lower than those for recombinant GluN2A, GluN2B, GluA1, or GluK2-containing receptors. It inhibits NMDA by blocking a conformational change necessary for channel opening and is non-competitive with co-agonists, glutamate and glycine.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Other Notes

Acker, T., et al. 2011. Mol. Pharmacol.80, 782.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service