Skip to Content
Merck
All Photos(1)

Documents

1.17988

Supelco

Flat-bottomed tubes with screw caps

for use with MQUANT®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

We offers you useful accessories. These flat-bottomed tubes can be used for our MQuant® test kits with color disk comparator (formally MColortest®). The screw caps are included.
for MQuant® tests with color disk comparator

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service