1.06828
Nicotinamide
test substance for elemental analysis
Synonym(s):
Nicotinamide, Nicotinamide, Niacinamide, 3-Pyridinecarboxamide
About This Item
Recommended Products
Quality Level
autoignition temp.
480 °C
potency
3530-3540 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)
pH
6.0-7.5 (20 °C, 50 g/L in H2O)
bp
150-160 °C/0.0007 hPa
mp
128-131 °C (sublimed)
transition temp
flash point 182 °C
solubility
691 g/L
density
1.40 g/cm3 at 20 °C
bulk density
360 kg/m3
storage temp.
15-25°C
InChI
1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)
InChI key
DFPAKSUCGFBDDF-UHFFFAOYSA-N
Analysis Note
Identity (IR-spectrum): conforms
C content (mean value tenfold determination): 58.7 - 59.3 %
H content (mean value tenfold determination): 4.8 - 5.2 %
N-content (mean value tenfold determination): 22.7 - 23.3 %
O content (mean value tenfold determination): 12.7 - 13.3 %
C content (standard deviation):
H content (standard deviation):
N-content (standard deviation):
O content (standard deviation):
Suitable as standard for the determination of C, H, N, O
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
302.0 °F - closed cup
Flash Point(C)
150 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service