Skip to Content
Merck
All Photos(1)

Key Documents

07-500

Sigma-Aldrich

Anti-p47-phox (mouse) Antibody

Upstate®, from rabbit

Synonym(s):

Cyclin-dependent kinase inhibitor p27, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CDKN1B(1027)

General description

Activation of NADPH-oxidase dependent superoxide generation requires the participation of several cytosolic factors including p47phox, p67phox, p40phox and Rac (1 or 2). In the resting cell, p47phox, p67phox, and p40phox form a large molecular weight complex which can be recovered from the cytosolic fraction. Activation of the NADPH oxidase is initiated by the assembly of p47phox, p67phox, and Rac with cytochrome b558 in a 1:1:1:1 complex in the plasma membrane. p47phox, p67phox, and Rac exhibit cooperative binding, since increasing the concentration of any one component lowers the EC50 of the other components. A great deal of current research involves understanding the protein protein interactions, and how these change with the activation state.

Specificity

Recognizes mouse p47-phox, MW ~47 kDa.

Immunogen

KLH-conjugated, synthetic peptides containing the first 15 amino acids of the N-terminal and the last 15 amino acids of the C-terminus of mouse p47-phox.

Application

Research Category
Neuroscience
Research Sub Category
Oxidative Stress
This Anti-p47-phox (mouse) Antibody is validated for use in WB for the detection of p47-phox (mouse).

Quality

Evaluated by western blot in RIPA lysates from RAW 264.7 cells.

Western Blot Analysis:
0.5-2.0 µg/mL of this antibody detected p47-phox in RIPA lysates from RAW 264.7 cells.

Target description

~47 kDa

Linkage

Replaces: 04-240

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, 0.05% sodium azide before the addition of glycerol to 30%.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
RAW 264.7 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janet E L Corry et al.
Journal of applied microbiology, 92(3), 424-432 (2002-03-02)
The prevalence and types of salmonella in broiler chickens during transportation and during slaughter and dressing were studied. This was part of a comprehensive investigation of salmonellas in two UK poultry companies, which aimed to find the origins and mechanisms
The redox-sensitive cation channel TRPM2 modulates phagocyte ROS production and inflammation.
Anke Di,Xiao-Pei Gao,Feng Qian,Takeshi Kawamura,Jin Han,Claudie Hecquet,Richard D Ye et al.
Nature Immunology null
J J Carrique-Mas et al.
Journal of applied microbiology, 107(6), 1976-1983 (2009-06-30)
To evaluate the performance of three Salmonella plating media (Rambach, Xylose Lysine Deoxycholate agar and modified Brilliant Green Agar plus Novobiocin) as part of the ISO 6579: 2002 (Annex D) on poultry environmental samples. The samples analysed were those for
Cloning and functional expression of the mouse homologue of p47phox.
S H Jackson et al.
Immunogenetics, 39(4), 272-275 (1994-01-01)
Laura J Norton et al.
Journal of cell science, 124(Pt 12), 1973-1983 (2011-05-26)
The signalling lipid phosphatidic acid (PA) is generated by the hydrolysis of phosphatidylcholine (PC), which is catalysed by phospholipase D (PLD) enzymes. Neutrophils, important cells of the innate immune system, maintain the body's defence against infection. Previous studies have implicated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service