Key Documents
Z255815
Sigma-Aldrich
Latex rubber laboratory tubing
I.D. × O.D. 1/4 in. × 3/8 in.
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
Pricing and availability is not currently available.
description
standard
packaging
pkg of 100 feet
I.D. × O.D.
1/4 in. × 3/8 in.
wall size
1/16 in.
General description
High-purity, seamless, amber tubing for general-purpose use with liquids and gases. Durometer: 35 ±5. Talc-free.
Choose from one of the most recent versions:
Certificates of Analysis (COA)