Skip to Content
Merck
All Photos(1)

Key Documents

W351407

Sigma-Aldrich

1,8-Octanedithiol

≥97%, FG

Synonym(s):

1,8-Dimercaptooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS(CH2)8SH
CAS Number:
Molecular Weight:
178.36
FEMA Number:
3514
EC Number:
Council of Europe no.:
2331
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.034
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥97%

refractive index

n20/D 1.505 (lit.)

bp

269-270 °C (lit.)

density

0.97 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty; sulfurous

SMILES string

SCCCCCCCCS

InChI

1S/C8H18S2/c9-7-5-3-1-2-4-6-8-10/h9-10H,1-8H2

InChI key

PGTWZHXOSWQKCY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,8-Octanedithiol is a volatile sulfur-containing flavoring agent with a meaty, sulfurous odor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of sulfur compounds in food flavor part III: Thiols.
Maga JA & Katz I.
Critical Reviews in Food Science and Nutrition, 7(2), 147-192 (1976)
Fenaroli's Handbook of Flavor Ingredients, 1523-1523 (2016)
Nidhi Shah et al.
ACS nano, 9(10), 10278-10286 (2015-09-08)
Here we describe the electrodeposition of Ag in the presence of cetyltrimethylammonium bromide (CTAB) onto 5 μm gap Au interdigitated array (IDA) electrodes that are bare, thiol-functionalized, or thiol-functionalized and seeded with 4 nm diameter Au nanoparticles (NPs). After deposition
Jin Yeong Na et al.
Scientific reports, 5, 13288-13288 (2015-08-25)
Spin-coating has been used extensively in the fabrication of electronic devices; however, the effects of the processing parameters have not been fully explored. Here, we systematically characterize the effects of the spin-coating time on the microstructure evolution during semiconducting polymer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service