Skip to Content
Merck
All Photos(1)

Documents

W208205

Sigma-Aldrich

Isoamyl propionate

≥98%, FCC, FG

Synonym(s):

Amyl propionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CO2CH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
144.21
FEMA Number:
2082
EC Number:
Council of Europe no.:
417
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.136
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.406 (lit.)

bp

156 °C (lit.)

density

0.871 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; banana; fruity; pineapple; sweet

SMILES string

CCC(=O)OCCC(C)C

InChI

1S/C8H16O2/c1-4-8(9)10-6-5-7(2)3/h7H,4-6H2,1-3H3

InChI key

XAOGXQMKWQFZEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Odor at 2.0%
Taste at 30 ppm

Other Notes

Natural occurrence: Fresh apple, tomato, white and red wine, apple wine.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 24, 51-51 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service