Skip to Content
Merck
All Photos(1)

Documents

MT51344761

Mettler Toledo Ionic Strength Adjuster

Calcium, perfectION ISE combination electrodes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115600
NACRES:
NB.79

form

liquid

manufacturer/tradename

Mettler-Toledo 51344761

volume

475 mL

Looking for similar products? Visit Product Comparison Guide

General description

Ionic strength adjuster (ISA) provides a constant background ionic strength for samples and standards.

Legal Information

perfectION is a trademark of Mettler-Toledo, Inc.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service