Skip to Content
Merck
All Photos(1)

Key Documents

M85409

Sigma-Aldrich

Methyl tridecanoate

98%

Synonym(s):

Tridecanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11COOCH3
CAS Number:
Molecular Weight:
228.37
Beilstein:
1769695
EC Number:
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.435 (lit.)

bp

131 °C/4 mmHg (lit.)

mp

5.5 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCC(=O)OC

InChI

1S/C14H28O2/c1-3-4-5-6-7-8-9-10-11-12-13-14(15)16-2/h3-13H2,1-2H3

InChI key

JNDDPBOKWCBQSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service