Skip to Content
Merck
All Photos(1)

Documents

901017

Sigma-Aldrich

N-Acetyl-Asp-Glu-13C5-OH

≥99 atom % 13C, ≥95% (CP)

Synonym(s):

Spaglumic acid-(glutamic acid-13C5)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C5C6H16N2O8
Molecular Weight:
309.22
UNSPSC Code:
12352106

isotopic purity

≥99 atom % 13C

Assay

≥95% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+5

storage temp.

−20°C

SMILES string

CC(N[C@@H](CC(O)=O)C(N[13C@@H]([13CH2][13CH2][13C](O)=O)[13C](O)=O)=O)=O

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service