Skip to Content
Merck
All Photos(2)

Key Documents

900963

Sigma-Aldrich

9-Phenyl-9′-(triphenylsilyl)-3,3′-bicarbazole

Synonym(s):

BCz-Si

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H34N2Si
CAS Number:
Molecular Weight:
666.88
UNSPSC Code:
12352005
NACRES:
NA.23

Assay

≥97%

form

powder

Orbital energy

HOMO 5.62 eV 
LUMO 2.3 eV 

Application

Blue phosphorescent OLED using BCz-Si as a host material achieved a max. Quantum efficiency of 21.0%, corresponding to a current efficiency and power efficiency as high as 46.5 cdA-1 and 45.8 lmW-1 respectively.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient blue/white phosphorescent organic light-emitting diodes based on a silicon-based host material via a direct carbon-nitrogen bond.
Xie YM, et al.
Journal of Material Chemistry C, 3(20), 5347-5353 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service