806013
APN-BCN
95%
Synonym(s):
Bicyclo[6.1.0]non-4-yn-9-ylmethyl (4-(cyanoethynyl)phenyl)carbamate
About This Item
Assay
95%
form
powder
reaction suitability
reagent type: cross-linking reagent
storage temp.
−20°C
SMILES string
N#CC#CC1=CC=C(NC(OCC2C3CCC#CCCC23)=O)C=C1
InChI
1S/C19H16N2O2/c20-13-5-6-14-9-11-15(12-10-14)21-19(22)23-18-16-7-3-1-2-4-8-17(16)18/h9-12,16-18H,3-4,7-8H2,(H,21,22)
InChI key
LVFLVSUFJDGHRN-UHFFFAOYSA-N
Application
Preparation Note
- Dissolve the protein in the appropriate buffer* with pH 6.5-9.0 (e.g. PBS) at 1-10 mg/mL concentration.
- Apply the appropriate amount of the stock solution of the reagent (1-5 molar eq. per free cysteine residue).
- Incubate at room temperature for 2 hours.
- If necessary; purify the protein-BCN conjugate using size exclusion chromatography or ultrafiltration.
- The conjugate can be readily coupled with azide-containing molecules by incubating the components in an aqueous buffer.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service