Skip to Content
Merck
All Photos(1)

Documents

792349

Sigma-Aldrich

MorDalphos Pd G2

Synonym(s):

Chloro(2-(di-1-adamantylphosphino)morpholinobenzene)[2-(2′-amino-1,1′-biphenyl)]palladium(II), MorDalphos-G2-palladacycle

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H52ClN2OPPd
CAS Number:
Molecular Weight:
773.72
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

feature

generation 2

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

>400 °C

functional group

phosphine

SMILES string

NC1=C(C=CC=C1)C2=C([Pd]Cl)C=CC=C2.P([C@]34C[C@@H](C[C@@H](C4)C5)C[C@@H]5C3)([C@@]6(C[C@@H](C7)C8)C[C@H]7C[C@H]8C6)C9=CC=CC=C9N%10CCOCC%10

InChI

1S/C30H42NOP.C12H10N.ClH.Pd/c1-2-4-28(27(3-1)31-5-7-32-8-6-31)33(29-15-21-9-22(16-29)11-23(10-21)17-29)30-18-24-12-25(19-30)14-26(13-24)20-30;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;;/h1-4,21-26H,5-20H2;1-6,8-9H,13H2;1H;/q;;;+1/p-1

InChI key

VEFSMOMAMGQMLF-UHFFFAOYSA-M

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service