75058
Tri-tert-butyl 1,4,7,10-tetraazacyclododecane-1,4,7-triacetate
≥97.0% (CHN)
Synonym(s):
DO3A-t-Bu-ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C26H50N4O6
CAS Number:
Molecular Weight:
514.70
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥97.0% (CHN)
form
lumps
impurities
≤4% water
SMILES string
CC(C)(C)OC(=O)CN1CCNCCN(CCN(CC1)CC(=O)OC(C)(C)C)CC(=O)OC(C)(C)C
InChI
1S/C26H50N4O6/c1-24(2,3)34-21(31)18-28-12-10-27-11-13-29(19-22(32)35-25(4,5)6)15-17-30(16-14-28)20-23(33)36-26(7,8)9/h27H,10-20H2,1-9H3
InChI key
NMHVTLJFPDOJOD-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jérôme Rossignol et al.
Sensors (Basel, Switzerland), 20(21) (2020-11-05)
A molecularly imprinted silica (MIS) coupled to a microwave sensor was used to detect three fungicides (iprodione, procymidone and pyrimethanil) present in most French wines. Chemometric methods were applied to interpret the microwave spectra and to correlate microwave signals and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service