435635
Cyclohexyl(dimethoxy)methylsilane
≥99%
Synonym(s):
Dow Corning® product Z-6187
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H11Si(OCH3)2CH3
CAS Number:
Molecular Weight:
188.34
Beilstein:
2827648
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Recommended Products
Assay
≥99%
form
liquid
refractive index
n20/D 1.439 (lit.)
bp
201.2 °C (lit.)
density
0.94 g/mL at 25 °C (lit.)
SMILES string
CO[Si](C)(OC)C1CCCCC1
Looking for similar products? Visit Product Comparison Guide
Legal Information
Repackaged by Aldrich Chemical Company, Inc. Manufactured by Dow Corning Corporation, U.S.A.
®Registered trademark of Dow Corning Corporation, U.S.A.
®Registered trademark of Dow Corning Corporation, U.S.A.
Dow Corning is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
150.8 °F - closed cup
Flash Point(C)
66 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service