Skip to Content
Merck
All Photos(1)

Key Documents

420425

Sigma-Aldrich

Phosphazene base P2-Et

98%

Synonym(s):

1-Ethyl-2,2,4,4,4-pentakis(dimethylamino)-2λ5,4λ5-catenadi(phosphazene), Tetramethyl(tris(dimethylamino)phosphoranylidene)phosphorictriamid-Et-imin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]3P=NP(=NC2H5)[N(CH3)2]2
CAS Number:
Molecular Weight:
339.40
Beilstein:
7485991
MDL number:
UNSPSC Code:
12352000

Assay

98%

refractive index

n20/D 1.492 (lit.)

bp

96 °C/0.05 mmHg (lit.)

density

1.02 g/mL at 25 °C (lit.)

SMILES string

CCN=P(N=P(N(C)C)(N(C)C)N(C)C)(N(C)C)N(C)C

InChI

1S/C12H35N7P2/c1-12-13-20(15(2)3,16(4)5)14-21(17(6)7,18(8)9)19(10)11/h12H2,1-11H3

InChI key

CFUKEHPEQCSIOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service