Skip to Content
Merck
All Photos(1)

Key Documents

360821

Sigma-Aldrich

(Z)-2-(2-Chloroacetamido)-α-(methoxyimino)-4-thiazoleacetic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8ClN3O4S
CAS Number:
Molecular Weight:
277.68
EC Number:
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

Assay

98%

SMILES string

CO\N=C(/C(O)=O)c1csc(NC(=O)CCl)n1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ochiai et al.
The Journal of antibiotics, 34(2), 160-170 (1981-02-01)
In an effort to improve the antibacterial activity of 7 beta-[2-(2-aminothiazol-4-yl)acetamido]-cephalosporins by introducing a methoxyimino group into the 7-acyl side chain, geometrically isomeric 2-(2-aminothiazol-4-yl)-2-methoxyiminoacetic acids and their derivatives were selectively synthesized. Structurally related acid derivatives were also synthesized. A facile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service