Skip to Content
Merck
All Photos(1)

Documents

345822

Sigma-Aldrich

2-Fluoro-3-(trifluoromethyl)aniline

97%

Synonym(s):

3-Amino-2-fluorobenzotrifluoride, α,α,α,2-Tetrafluoro-m-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CF3)NH2
CAS Number:
Molecular Weight:
179.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.462 (lit.)

bp

189 °C (lit.)

density

1.39 g/mL at 25 °C (lit.)

SMILES string

Nc1cccc(c1F)C(F)(F)F

InChI

1S/C7H5F4N/c8-6-4(7(9,10)11)2-1-3-5(6)12/h1-3H,12H2

InChI key

YKPDYPPZLUZONK-UHFFFAOYSA-N

Application

2-Fluoro-3-(trifluoromethyl)aniline may be used in chemical synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service