Skip to Content
Merck
All Photos(1)

Key Documents

343889

Sigma-Aldrich

1,2-Bis(2-Aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid

technical grade, 90%

Synonym(s):

2,2′-(Ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid, BAPTA, Ethylenedioxybis(o-phenylenenitrilo)tetraacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O10
CAS Number:
Molecular Weight:
476.43
Beilstein:
5192406
MDL number:
Pricing and availability is not currently available.

grade

technical grade

Assay

90%

SMILES string

OC(=O)CN(CC(O)=O)c1ccccc1OCCOc2ccccc2N(CC(O)=O)CC(O)=O

InChI

1S/C22H24N2O10/c25-19(26)11-23(12-20(27)28)15-5-1-3-7-17(15)33-9-10-34-18-8-4-2-6-16(18)24(13-21(29)30)14-22(31)32/h1-8H,9-14H2,(H,25,26)(H,27,28)(H,29,30)(H,31,32)

InChI key

FTEDXVNDVHYDQW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shan Li et al.
Molecular cell, 74(6), 1123-1137 (2019-05-06)
Abnormal processing of stressed replication forks by nucleases can cause fork collapse, genomic instability, and cell death. Despite its importance, it is poorly understood how the cell properly controls nucleases to prevent detrimental fork processing. Here, we report a signaling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service