167428
(2-Carbamoylphenoxy)acetic acid sodium salt
97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NH2C(O)C6H4OCH2CO2Na
CAS Number:
Molecular Weight:
217.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
97%
mp
267 °C (lit.)
SMILES string
[Na+].NC(=O)c1ccccc1OCC([O-])=O
InChI
1S/C9H9NO4.Na/c10-9(13)6-3-1-2-4-7(6)14-5-8(11)12;/h1-4H,5H2,(H2,10,13)(H,11,12);/q;+1/p-1
InChI key
OEQSACUVTKYXIR-UHFFFAOYSA-M
General description
(2-Carbamoylphenoxy)acetic acid is commonly used as analgesic.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The synthesis and characterisation of some ortho-substituted phenoxyalkanoic acids and their derivatives.
Lupea AX, et al.
J. Serb. Chem. Soc., 71(12), 1247-1261 (2006)
D Loew et al.
International journal of clinical pharmacology, therapy, and toxicology, 29(3), 124-129 (1991-03-01)
The pharmacokinetics of o-carbamoylphenoxyacetic acid was studied in 9 male healthy subjects following a single i.m. administration of 1000 mg of this compound in form of its sodium salt. Venous blood specimens were drawn up to the 10th h p.a.
Pharmacokinetics and efficacy of carbamoylphenoxyacetid acid.
D Loew et al.
International journal of clinical pharmacology, therapy, and toxicology, 30(11), 509-511 (1992-11-01)
[Theophylline solubilizer and salicylate asthma].
K Drebinger et al.
Fortschritte der Medizin, 104(23-24), 480-482 (1986-06-19)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service