Skip to Content
Merck
All Photos(2)

Key Documents

SAB5201790

Sigma-Aldrich

Monoclonal Anti-ENaC beta-Atto 488 antibody produced in mouse

clone 7B8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
conjugate:
Atto 488 conjugate
application:
IHC
WB
clone:
7B8, monoclonal
species reactivity:
mouse
citations:
technique(s):
immunohistochemistry: 1:150
western blot: 1:1000

biological source

mouse

conjugate

Atto 488 conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7B8, monoclonal

form

buffered aqueous solution

mol wt

87kDa

species reactivity

mouse

concentration

1.0 mg/mL

technique(s)

immunohistochemistry: 1:150
western blot: 1:1000

isotype

IgG1

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Synthetic peptide from the C-terminal of Rat ENaC beta (aa. 617-638)

Physical form

PBS pH 7.4, 50% glyercol, 0.09% Sodium Azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service