Skip to Content
Merck
All Photos(1)

Key Documents

EHU005381

Sigma-Aldrich

MISSION® esiRNA

targeting human MDM4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGGGACGTCAGAGCTTCTCCGTGAAAGACCCAAGCCCTCTCTATGATATGCTAAGAAAGAATCTTGTCACTTTAGCCACTGCTACTACAGATGCTGCTCAGACTCTCGCTCTCGCACAGGATCACAGTATGGATATTCCAAGTCAAGACCAACTGAAGCAAAGTGCAGAGGAAAGTTCCACTTCCAGAAAAAGAACTACAGAAGACGATATCCCCACACTGCCTACCTCAGAGCATAAATGCATACATTCTAGAGAAGATGAAGACTTAATTGAAAATTTAGCCCAAGATGAAACATCTAGGCTGGACCTTGGATTTGAGGAGTGGGATGTAGCTGGCCTGCCTTGGTGGTTTTTAGGAAACTTGAGAAGCAACTATACACCTAGAAGTAATGGCTCAACTGATTTACAGACAAATCAGGATGTGGGTACTGCCATT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haishan Zhang et al.
Molecular immunology, 125, 9-14 (2020-07-04)
MiR-483-3p is involved in the pathogenesis of acute myocardial infarctions, but its association with myocardial ischemia reperfusion (IR) remains mostly unknown. In this study, an in vitro model of myocardial IR injury was established by putting H9c2 cells into hypoxia
Kehua Jiang et al.
Molecular genetics & genomic medicine, 7(8), e833-e833 (2019-06-30)
MicroRNA-33a (miR-33a) plays the role of the tumor suppressor gene by regulating the expression level of downstream genes. However, the effects of miR-33a in renal cell cancer (RCC) remain unknown. Our study was designed to investigate the expression level and
Hong Yan et al.
American journal of cancer research, 9(2), 312-329 (2019-03-25)
Activated KRAS is frequently observed and paralleled by inactivating of tumor suppressors in lung cancer, while the mechanisms remained elusive. Here, our study revealed a microRNA was involved in KRAS overexpression, activation of KRAS signaling and its synergy with inactivating
Yan Li et al.
Journal of molecular histology, 46(4-5), 357-364 (2015-06-21)
Multidrug resistance-associated protein 1 (MRP1) belongs to ATP-binding cassette transporters family. The overexpression of MRP1 is predominantly related with the failure of chemo-radiotherapy in various tumors. However, its possible role in hypertrophic scar (HS) is hardly investigated. Here we showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service