Skip to Content
Merck
All Photos(1)

Documents

D2922000

Disopyramide phosphate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Disopyramide phosphate salt, α-Diisopropylaminoethyl-α-phenylpyridine-2-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H29N3O · H3PO4
CAS Number:
Molecular Weight:
437.47
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

disopyramide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OP(O)(O)=O.CC(C)N(CCC(C(N)=O)(c1ccccc1)c2ccccn2)C(C)C

InChI

1S/C21H29N3O.H3O4P/c1-16(2)24(17(3)4)15-13-21(20(22)25,18-10-6-5-7-11-18)19-12-8-9-14-23-19;1-5(2,3)4/h5-12,14,16-17H,13,15H2,1-4H3,(H2,22,25);(H3,1,2,3,4)

InChI key

CGDDQFMPGMYYQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Disopyramide phosphate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Class IA antiarrhythmic; sodium channel blocker

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Disopyramide phosphate: the electrophysiological mechanisms of its anti-arrhythmia action and its use in clinical practice].
A A Grosu
Kardiologiia, 25(5), 114-118 (1985-05-01)
R Di Bianco et al.
Angiology, 38(2 Pt 2), 174-183 (1987-02-01)
In the absence of preexistent myocardial dysfunction, the risk of producing cardiac decompensation in patients being treated with any of the conventional antiarrhythmic agents is low. The availability of disopyramide for clinical use in 1977 produced optimism for an effective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service