Skip to Content
Merck
All Photos(1)

Documents

32522

Supelco

Cyprosulfamide

PESTANAL®, analytical standard

Synonym(s):

N-[4-(Cyclopropylaminocarbonyl)phenylsulfonyl]-2-methoxybenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N2O5S
CAS Number:
Molecular Weight:
374.41
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1ccccc1C(=O)NS(=O)(=O)c2ccc(cc2)C(=O)NC3CC3

InChI

1S/C18H18N2O5S/c1-25-16-5-3-2-4-15(16)18(22)20-26(23,24)14-10-6-12(7-11-14)17(21)19-13-8-9-13/h2-7,10-11,13H,8-9H2,1H3,(H,19,21)(H,20,22)

InChI key

OAWUUPVZMNKZRY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyprosulfamide is a herbicide, and can protect the plants from salinity stress and induced vigorous growth, including the formation of new tillers and early flowering.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

George Giannakopoulos et al.
Pest management science, 76(10), 3413-3422 (2020-02-23)
Safeners extend the application of existing herbicides by selectively enhancing tolerance in large-grained cereal crops. While their activity is linked to enhanced herbicide metabolism, their exact mode of action and reasons for their crop specificity have yet to be determined.
Abscisic acid and the herbicide safener cyprosulfamide cooperatively enhance abiotic stress tolerance in rice
Dashevskaya, Svetlana, et al.
Molecular breeding : new strategies in plant improvement, 32 (2), 463-484 (2013)
Amit Paporisch et al.
Pesticide biochemistry and physiology, 138, 22-28 (2017-05-01)
Three sweet corn genotypes, two inbred lines (IBER001 and IBER002) and their hybrid (ER00X), differ in their phenotypic responses to several P450-metabolized herbicides, used in sweet corn, namely, foramsulfuron, iodosulfuron, rimsulfuron and tembotrione. Foramsulfuron is a sulfonylurea herbicide commonly formulated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service