Z219800
Single-bulb ball-tube
capacity 250 mL, joint: ST/NS 24/40, w/drip-tip
All Photos(2)
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
Joint
joint: ST/NS24/40
capacity
250 mL
Related Categories
General description
Arranged horizontally outside of Kugelrohr oven, the ball-tubes cool and collect distillate with minimal loss of material.
Choose from one of the most recent versions:
Certificates of Analysis (COA)