Key Documents
Z744067
FoilSeal Extra Thick Sealing Foil
aluminum, non-sterile
About This Item
UNSPSC Code:
41122410
NACRES:
NB.04
material
aluminum
sterility
non-sterile
feature
RNase and DNase free
packaging
case of 100 ea
manufacturer/tradename
Excel Scientific F2-100
parameter
-40 °C temp. range (-20 °C)
W × L
78 mm × 128 mm
General description
FoilSeal sealing foils remove cleanly from plate surfaces with no adhesive stringing, residue or contamination. Each sealing foil measures 78.3 x 128 mm and offers sufficient sealing area for all plate types (e.g. flat top and raised-rim). Chamfered corners result in no plate overhang for robotics applications. Features one end-tab with perforations. Length between the perforation with end tab removed and the end of the foil seal is 118.1 mm.
  • Universal fit for use with all plate types (e.g. flat top and raised-rim)
  • The smaller dimensions and chamfered corners result in no plate overhang for robotics applications
  • Single end-tab features a custom perforation that will not tear during removal
  • Certified DNase- and RNase- free
  • Excellent barrier properties minimizes sample evaporation
  • Conformable foil allows visual verification of seal security
  • Pierceable with pipet tips and robotic probes for direct sample retrieval
Legal Information
FoilSeal is a trademark of Excel Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)