Skip to Content
Merck
All Photos(1)

Key Documents

NIST1957

Organic contaminants in non-fortified human serum

NIST® SRM® 1957

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10.7 mL reconstituted

manufacturer/tradename

NIST®

application(s)

clinical research

format

matrix material

General description

This Standard Reference Material (SRM) is intended for use in evaluating analytical methods for the determination of selected polychlorinated biphenyl (PCB) congeners, chlorinated pesticides, and polybrominated diphenyl ether (PBDE) congeners in human serum and similar matrices. A unit of SRM 1957 consists of five 30 mL vials each containing freeze-dried non-fortified human serum. For more information, please refer to the COA and SDS.

SRM 1957_cert

SRM 1957_SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service