Skip to Content
Merck
All Photos(1)

Key Documents

8.22281

Sigma-Aldrich

Indole

for synthesis

Synonym(s):

Indole, 2,3-Benzopyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N
CAS Number:
Molecular Weight:
117.15
MDL number:
UNSPSC Code:
12352005
EC Index Number:
204-420-7
NACRES:
NA.22

vapor pressure

0.016 hPa ( 25 °C)

Quality Level

Assay

≥99.0% (GC)

form

solid

potency

1000 mg/kg LD50, oral (Rat)
790 mg/kg LD50, skin (Rabbit)

pH

5.9 (20 °C, 1000 g/L in H2O)

bp

253 °C/1013 hPa

mp

52 °C

transition temp

flash point 110 °C

density

1.22 g/cm3 at 20 °C

bulk density

230 kg/m3

storage temp.

2-30°C

InChI

1S/C8H7N/c1-2-4-8-7(3-1)5-6-9-8/h1-6,9H

InChI key

SIKJAQJRHWYJAI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 50 °C
Melting range (upper value): ≤ 52 °C
Identity (IR): passes test

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service