Key Documents
BAH500250409
SP Bel-Art® Glove Box Sleeved Gloves for 8 in. Glove Ports
size 9 medium
Synonym(s):
glove box
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
Pricing and availability is not currently available.
material
black neoprene
size
9 medium
packaging
pkg of 1 pair
manufacturer/tradename
Bel-Art H50025-0409
General description
One-piece, seamless, smooth neoprene protection against liquid, gas or solid substance penetration.
Features and Benefits
  • Suitable for use with glove boxes having 8" glove ports including sid ENTRY H50026-0000 Glove Boxes
  • Requires clamping rings (H50025-0320) to hold gloves in place
  • Sold in pairs
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)