Apoptosis Assays
Cell Based Assays for Apoptosis Detection
Apoptosis, or programmed cell death, is a growth-limiting regulatory mechanism by which cells can trigger their own death in response to extracellular signals because of irreparable cellular or DNA damage. The ability of tumor cells to elude apoptosis is a hallmark of most types of cancer. Apoptosis also plays an important role in developmental mechanisms, such as preventing the overgrowth of neuronal cell lineages in the developing brain and regulating interdigital spacing in limb development. Autophagy and apoptosis are connected and extensive crosstalk occurs between the two. For example, during nutrient deficiency, autophagy functions as a pro-survival mechanism. However, excessive autophagy may lead to cell death, a process morphologically distinct from apoptosis. Apoptosis is a multistep process including early, mid and late-stage cellular events which can be detected using various cellular assays including Annexin V, Caspase and TUNEL detection methods outlined below.
Stages of Apoptosis
Early-Stage Events: Translocation of phosphatidylserine to the outer leaf of plasma membrane (1), loss of mitochondrial membrane potential (2), cytochrome C & ATP release (3) and activation of caspase-8 and 9 (4).
Mid-Stage Events: Activation of caspase-3, 6 and 7 (5), Poly-ADP-Ribose polymerase (PARP) cleavage (6), cell shrinkage (7), and activation of nucleases (8).
Late-Stage Events: DNA fragmentation (9), nuclear collapse (10), formation of apoptotic bodies (11), and phagocytosis by macrophages (12).
Early-Stage Apoptosis
Annexin V Assays: Phosphatidylserine is normally confined in the inner membrane leaflet of viable cells. The translocation of phosphatidylserine to the exposed membrane surface is an early event in apoptosis, where it serves as a signal for the attack of phagocytic cells. Annexins are a family of structurally related proteins that can bind specifically to cellular membranes. Annexin V has a very high affinity for membranes containing the negatively charged phospholipid phosphatidylserine and can be used as marker of early apoptosis events.
Product Description | Detection Options | Sample Type | Application | Product Number |
---|---|---|---|---|
ApopNexin™ Kits (EMD) | Fluorometric | Adherent Cells/ Suspension Cells | Immunofluorescence | APT750, PF032, CBA059, CBA060 |
Guava/Muse Annexin V Assay Kits (EMD) | Fluorometric | Suspension Cells | Flow Cytometry | 4500-0450, 4500-0455, FCCH100108, MCH100105 |
Annexin-V Kits (Sigma) | Fluorometric | Adherent Cells/ Suspension Cells | Immunofluorescence Flow Cytometry | APOAF, APOAC, ROANNV, 11828681001 |
Figure 1.Dot plots depicting Jurkat cells treated with various concentrations of apoptosis inducers and stained using Annexin Red Kit (FCCH100108). Jurkat cells were untreated (Plot A), or treated with 0.1 uM (Plot B) or with 3 uM Staurosporine (Plot C) and then stained using FlowCelllect Annexin Red Kit.
Mitochondrial Detection: Mitochondria are important cellular organelles that maintain crucial cellular energy balance, contain key regulators of cell death processes such as apoptosis and play a significant role in cellular oxidative stress generation. Collapse of mitochondrial membrane potential is believed to coincide with the permeabilization of the outer mitochondrial membrane, and release of Cytochrome C and other pro-apoptotic proteins into the cytosol, which then triggers the downstream events in the apoptotic cascade.
Product Description | Detection Options | Sample Type | Application | Product Number |
---|---|---|---|---|
Guava® Mitochondrial Depolarization/Stress/ Damage Kits (EMD) | Fluorometric | Suspension Cells | Flow Cytometry | 4500-0250, FCCH100109, FCCH100106 |
MitoLight® Mitochondrial Apoptosis Detection Kits (EMD) | Fluorometric | Adherent Cells/ Suspension Cells | Immunofluorescence | APT142, APT242 |
BioTracker Mitochondria Dyes (EMD) | Fluorometric | Adherent Cells/ Suspension Cells | Live Cell Imaging Flow Cytometry | SCT135, SCT136, SCT137 |
Mitochondrial Membrane Potential Kit (Sigma) | Fluorometric | Adherent Cells/ Suspension Cells | Immunofluorescence/ Flow Cytometry | MAK159, MAK160 |
Cytochrome C Assay Kits (Sigma) | Colorimetric | Adherent Cells/ Suspension Cells | Plate Reader | CY0100, CYTOCOX1 |
Mitochondria Isolation Kits (Sigma) | N/A | Adherent Cells/ Suspension Cells | Isolation | MITOISO1, MITOISO2, CS0760 |
JC-1 Dye (Sigma) | Fluorometric | Adherent Cells/ Suspension Cells | Live Cell Imaging | T4069 |
Mid-Stage Apoptosis
Caspase Detection Assays: A central component of the apoptotic process is a cascade of proteolytic enzymes called caspases. Caspases participate in a series of reactions that are triggered in response to pro-apoptotic signals and result in the cleavage of protein substrates, causing the disassembly of the cell. Caspase enzymes specifically recognize a 4 or 5 amino acid sequence on the target substrate which necessarily includes an aspartic acid residue. This residue is the target for cleavage, which occurs at the carbonyl end of the aspartic acid residue.
Live Cell Imaging of Apoptosis in Immune Cells using the CellASIC® ONIX2 Microfluidic System
Product Description | Detection Options | Sample Type | Application | Product Number |
---|---|---|---|---|
BioTracker NucView® Caspase-3 Dyes (EMD) | Fluorometric | Adherent Cells/ Suspension Cells | Live Cell Imaging Flow Cytometry | SCT100, SCT101, SCT102, SCT103, SCT104, SCT105 |
CaspaTag™ Pan-Caspase Caspase-3,7 Caspase-8 Caspase-9 (EMD) | Fluorometric | Adherent Cells/ Suspension Cells/Tissue | Immunofluorescence/ Cytometry/Plate Reader | APT400, APT420, APT403, APT423, APT408, APT428, APT409, APT429 |
CaspSCREEN™ Kit (EMD) | Fluorometric | Suspension Cells | Cytometry | APT105 |
Guava® Caspase Kits (EMD) | Fluorometric | Suspension Cells | Flow Cytometry | 4500-0500, 4500-0530, 4500-0540, 4500-0550 |
Caspase Activity Kits (EMD) | Fluorometric/ Colorimetric/ Chemiluminescent | Cell Lysates | Plate Reader | APT129, APT131, APT165, APT171, APT173 |
Caspase 3/8 Assay Kit (Sigma) | Fluorometric/ Colorimetric | Adherent Cells/ Suspension Cells | Immunofluorescence/ Cytometry | CASP3C, CASP3F, CASP8C, APPA015 |
Figure 2. Live cell detection of mitochondrial membrane potential, caspase activity, and apoptosis in MCF-7 breast cancer cells.Untreated MCF-7 cells (86012803) (A) or treated with staurosporine (S5921) overnight to induce apoptosis (B), then stained with the BioTracker™ 405 Blue Mitochondria Dye (SCT135), BioTracker NucView® 530 Red Caspase-3 Dye (SCT105), and CF®488A Annexin V for 30 minutes at 37°C in cell culture medium with no wash. Healthy cells show mitochondrial staining (cyan), apoptotic cells lose blue staining (cyan), and show increased staining of nuclei (red) and CF®488A Annexin V staining of cellular membranes (green).
Late-Stage Apoptosis
TUNEL Assays: DNA fragmentation in apoptosis is usually associated with structural changes in cellular morphology and is a hallmark of late-stage apoptosis. DNA fragmentation in apoptosis can be examined using the TUNEL assay. The in situ staining of DNA strand breaks detected by the TUNEL assay and subsequent visualization by light microscopy gives biologically significant data about DNA damage and late-stage apoptosis.
Figure 3.The ApopTag™ family of apoptosis kits examine DNA fragmentation by the TUNEL assay. The DNA strand breaks are detected by enzymatically labeling the free 3’-OH termini with modified nucleotides. These new DNA ends that are generated upon DNA fragmentation are typically localized in morphologically identifiable nuclei and apoptotic bodies.
Product Description | Detection Options | Sample Type | Application | Product Number |
---|---|---|---|---|
Guava® TUNEL Kit (EMD) | Fluorometric | Suspension cells | Flow Cytometry | 4500-0121 |
ApopTag® TUNEL Kits (EMD) | Fluorometric/ Colorimetric | Adherent Cells/ Suspension Cells/Tissue | Immunohistochemistry/ Immunofluorescence/ Cytometry | S7100, S7101, S7110, S7111, S7160, S7165 |
ApopTag® ISOL Kits (EMD) | Fluorometric/ Colorimetric | Adherent Cells/ Suspension Cells/Tissue | Immunohistochemistry/ Immunofluorescence | APT1000, S7200 |
FragEL™ DNA Fragmentation Detection Kits (EMD) | Fluorometric/ Colorimetric | Adherent Cells/ Suspension Cells/Tissue | Immunohistochemistry/ Immunofluorescence | QIA21, QIA33, QIA39 |
Flow Cytometry Kit for Apoptosis (Sigma) | Fluorometric | Suspension cells | Flow Cytometry | APOBRDU |
Cell Death Detection Kits (Roche) | Fluorometric/ Colorimetric | Adherent Cells/ Suspension Cells/ Tissue | Immunohistochemistry/ Immunofluorescence/ ELISA | 06432344001, 11835246001, 11585045001, 11684795910, 11684809910, 11684817910, 12156792910, CELLDETH-RO |
Figure 4.A) Detection of apoptotic cells in human lymph node with ApopTag® Peroxidase (S7200). The tissue was fixed in 10% neutral-buffered formalin and paraffin-embedded. B) Detection of apoptotic cells with ApopTag® Fluorescein Direct kit (S7160) in mouse embryo forelimb bud, fixed with 10% neutral-buffered formalin and paraffin-embedded. C) Detection of apoptotic cells with ApopTag® Red (S7165) in human lymph node tissue, fixed with 10% neutral-buffered formalin and paraffin-embedded.
H2A.X Assays: Double-stranded DNA breaks (DSB) are generated that rapidly result in the phosphorylation of histone variant H2A.X. Because phosphorylation of H2A.X at Ser139 correlates well with each DSB, phospho-H2A.X is a sensitive marker used to examine DNA damage and late stage apoptosis.
Product Description | Detection Options | Sample Type | Application | Product Number |
---|---|---|---|---|
H2A.X Phosphorylation Assay Kit (EMD) | Chemiluminescent | Adherent Cells/ Suspension Cells | ELISA | 17-327 |
H2A.X Phosphorylation Assay Kit (EMD) | Fluorometric | Suspension cells | Flow Cytometry | 17-344 |
FlowCellect™ Histone H2A.X Phosphorylation Assay Kit (EMD) | Fluorometric | Suspension cells | Flow Cytometry | FCCS100182 |
Conclusion
Programmed cell death (apoptosis) is controlled by multiple signaling and effector pathways linked to mitochondrial depolarization, caspase related apoptotic enzyme cascades, DNA fragmentation and ultimately cell blebbing and destruction. The integrity of these and other links can be genetically compromised in many diseases such as cancer making detection and measurement of apoptosis extremely useful in many research assays. Our well-published apoptosis assays, including the ApopTag® family of the TUNEL assays and the CaspaTag™ family of In Situ Caspase detection assays allows reproducible insights into cell death mechanisms.