Z742093
Western Blot Box
size (square), size 11.4 cm × 11.4 cm × 3.2 cm, pk of 5
About This Item
Código UNSPSC:
41105300
NACRES:
NB.22
Productos recomendados
tamaño
(square)
11.4 cm × 11.4 cm × 3.2 cm
envase
pk of 5
color
black
Descripción general
- Perfect box for every blot
- Reduce antibody usage
- Hinged Lid minimizes evaporation
- For halves, wholes, strips, or minis
No matter what size your Western blot is, there′s a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at 90° angles and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies (for example, popular mini-gels require only about 4mL).
At a price that makes them truly disposable, all sizes are engineered from crystal clear polystyrene. All sizes are also equipped with an easy-to-open hinged lid that reduces evaporation.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)