Documentos clave
Z332461
BRAND® discharge tube for Dispensette®
with integrated valve, fine tip, for 25, 50 and 100 mL Dispensette III
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
manufacturer/tradename
BRAND
L
120 mm
color
red
General description
The BRAND® Discharge Tube is designed for use with both the Dispensette® and Seripettor® systems, providing a reliable solution for precise liquid dispensing. This versatile discharge tube includes essential components such as a closure cap, EPDM discharge valve, and sealing ring, ensuring optimal performance and convenience.
Other Notes
BR707918 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG