Documentos
Z290637
Sigma-Aldrich
Vacuum filtration assembly flasks
collection flask, capacity 4000 mL
About This Item
UNSPSC Code:
41121804
NACRES:
NB.24
material
borosilicate glass
glass housing
sterility
non-sterile
packaging
pack of 1
pkg of 1 ea
manufacturer/tradename
Sigma-Aldrich
bottle capacity
4000 mL
capacity
4000 mL
funnel capacity
4000 mL
fitting
connector for 40/35 joint
General description
Flask does not have side arm.