HS120258
Modular glove box holder
W × L × H 254 mm × 156 mm × 97 mm, white
About This Item
Código UNSPSC:
42132201
NACRES:
NB.17
Productos recomendados
Materiales
plastic
Características
Holds 1 box
fabricante / nombre comercial
Heathrow Scientific HS120258
An × L × Al
254 mm × 156 mm × 97 mm
color
white
Descripción general
Available infused with Microban ideal for use in clean rooms, hospitals, and microbiological work areas.
- High-impact, durable resin is resistant to harsh cleaning solutions
- Modular design allows single boxes or horizontal stacking for multiple box holder
- Universal fit for most major brands of glove boxes
- Mounting screws included
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)