Documentos clave
EP5910755007
Adapter for Eppendorf® S-4x Universal Large Rotor
13x75-100 mm round-bottom rubes, pkg of 2 ea
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
13x75-100 mm round-bottom rubes
Holds 4 - 8 mL tubes
packaging
pkg of 2 ea
manufacturer/tradename
Eppendorf® 5910755007
technique(s)
centrifugation: suitable
compatibility
for use with Universal bucket
General description
Adapters for various sizes of tubes and bottles for use with Eppendorf S-4x Universal large rotor.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG