Merck
Todas las fotos(4)
Documentos clave
EP4925000111
Eppendorf® Reference® 2 Fixed Volume Pipettor
single-channel, volume 200 μL, yellow
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
Materiales
yellow
Características
single-channel
mechanical
envase
pack of 1 ea
fabricante / nombre comercial
Eppendorf® 4921000117 (Global)
volumen
200 μL
Descripción general
The Reference 2 is the successor to the highly successful Reference pipette, and enhances the range′s already high standards with a new design, reduced weight and operating forces and multi-channel versions. As the premium product in Eppendorf′s pipette portfolio, the Reference 2 provides the most accurate results possible alongside robust, reliable handling and optimum user safety.
These levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducability, Eppendorf′s new Reference 2 is an ideal instrument for these liquid handling applications.
Información legal
Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)